Assay Details
| Gene Symbol: RBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202939.1 | NP_001189868.1 | 4 - 5 | 623 | 110 | |
| NM_112064.5 | NP_566417.3 | 4 - 5 | 812 | 110 | ||
| GenBank mRNA | AF245395.1 | - | 4 - 5 | 530 | 110 | |
| AY150438.1 | - | 3 - 4 | 326 | 110 | ||
More Information
Set Membership: |
|

RBR1
Genomic Map