Assay Details
| Gene Symbol: RBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202939.1 | NP_001189868.1 | 6 - 7 | 767 | 64 | |
| NM_112064.5 | NP_566417.3 | 6 - 7 | 956 | 64 | ||
| GenBank mRNA | AF245395.1 | - | 6 - 7 | 674 | 64 | |
More Information
Set Membership: |
|

RBR1
Genomic Map