Assay Details
| Gene Symbol: BUG22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112066.5 | NP_566418.1 | 1 - 2 | 119 | 91 | |
| GenBank mRNA | AY057521.1 | - | 1 - 2 | 85 | 91 | |
| AY113056.1 | - | 1 - 2 | 56 | 91 | ||
More Information
Set Membership: |
|

BUG22
Genomic Map