Assay Details
| Gene Symbol: MGT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119000.3 | NP_194587.2 | 1 - 2 | 288 | 118 | |
| GenBank mRNA | DQ446879.1 | - | 1 - 2 | 288 | 118 | |
More Information
Set Membership: |
|

MGT5
Genomic Map