Assay Details
| Gene Symbol: PHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119003.4 | NP_194590.2 | 2 - 3 | 747 | 124 | |
| GenBank mRNA | AJ310799.1 | - | 2 - 3 | 657 | 124 | |
| AY081290.1 | - | 2 - 3 | 714 | 124 | ||
| BT002187.1 | - | 1 - 2 | 559 | 124 | ||
More Information
Set Membership: |
|

PHR1
Genomic Map