Assay Details
| Gene Symbol: PHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119003.4 | NP_194590.2 | 7 - 8 | 1189 | 72 | |
| GenBank mRNA | AJ310799.1 | - | 7 - 8 | 1099 | 72 | |
| AY081290.1 | - | 7 - 8 | 1156 | 72 | ||
| BT002187.1 | - | 6 - 7 | 1001 | 72 | ||
More Information
Set Membership: |
|

PHR1
Genomic Map