Assay Details
| Gene Symbol: TYRDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084992.1 | NP_001078461.1 | 6 - 7 | 962 | 133 | |
| NM_001203932.1 | NP_001190861.1 | 6 - 7 | 1096 | 133 | ||
| NM_001203933.2 | NP_001190862.1 | 7 - 8 | 1049 | 133 | ||
| NM_001341927.1 | NP_001329473.1 | 7 - 8 | 1061 | 133 | ||
| NM_001341928.1 | NP_001329474.1 | 7 - 8 | 1049 | 133 | ||
| NM_001341929.1 | NP_001329472.1 | 7 - 8 | 1049 | 133 | ||
| NM_119010.3 | NP_194597.1 | 6 - 7 | 962 | 133 | ||
More Information
Set Membership: |
|

TYRDC
Genomic Map