Assay Details
| Gene Symbol: GrxC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119017.5 | NP_194602.2 | 4 - 5 | 574 | 75 | |
| GenBank mRNA | AK118672.1 | - | 4 - 5 | 521 | 75 | |
| BT003703.1 | - | 4 - 5 | 443 | 75 | ||
More Information
Set Membership: |
|

GrxC5
Genomic Map