Assay Details
| Gene Symbol: ROP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119039.3 | NP_194624.1 | 4 - 5 | 727 | 67 | |
| GenBank mRNA | AF079484.1 | - | 4 - 5 | 524 | 67 | |
| BT025271.1 | - | 4 - 5 | 360 | 67 | ||
More Information
Set Membership: |
|

ROP9
Genomic Map