Assay Details
| Gene Symbol: AT4G29000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119044.2 | NP_194629.1 | 3 - 4 | 1051 | 90 | |
| GenBank mRNA | AK118658.1 | - | 3 - 4 | 1051 | 90 | |
| BT005931.1 | - | 3 - 4 | 607 | 90 | ||
More Information
Set Membership: |
|

AT4G29000
Genomic Map