Assay Details
| Gene Symbol: emb2726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119050.3 | NP_567820.1 | 5 - 6 | 2514 | 63 | |
| GenBank mRNA | AY056792.1 | - | 5 - 6 | 2458 | 63 | |
More Information
Set Membership: |
|

emb2726
Genomic Map