Assay Details
| Gene Symbol: SNF7.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084999.2 | NP_001078468.1 | 6 - 7 | 963 | 97 | |
| NM_119060.5 | NP_194645.1 | 7 - 8 | 926 | 97 | ||
| NM_202906.1 | NP_974635.1 | 5 - 6 | 1121 | 97 | ||
| GenBank mRNA | AY042853.1 | - | 7 - 8 | 761 | 97 | |
| AY072490.1 | - | 6 - 7 | 656 | 97 | ||
More Information
Set Membership: |
|

SNF7.1
Genomic Map