Assay Details
| Gene Symbol: PFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119080.4 | NP_194664.1 | 1 - 2 | 370 | 79 | |
| GenBank mRNA | AK319124.1 | - | 2 - 3 | 192 | 79 | |
| AY074350.1 | - | 1 - 2 | 216 | 79 | ||
| AY114048.1 | - | 1 - 2 | 123 | 79 | ||
| U43326.1 | - | 1 - 2 | 166 | 79 | ||
More Information
Set Membership: |
|

PFN2
Genomic Map