Assay Details
| Gene Symbol: NFC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119118.3 | NP_194702.2 | 13 - 14 | 1341 | 70 | |
| GenBank mRNA | AK176425.1 | - | 13 - 14 | 1278 | 70 | |
| BT010610.1 | - | 13 - 14 | 1262 | 70 | ||
More Information
Set Membership: |
|

NFC5
Genomic Map