Assay Details
| Gene Symbol: NFC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341975.1 | NP_001329130.1 | 7 - 8 | 719 | 92 | |
| NM_001341976.1 | NP_001329131.1 | 7 - 8 | 707 | 92 | ||
| NM_119118.3 | NP_194702.2 | 7 - 8 | 729 | 92 | ||
| GenBank mRNA | AK176425.1 | - | 7 - 8 | 666 | 92 | |
| BT010610.1 | - | 7 - 8 | 650 | 92 | ||
More Information
Set Membership: |
|

NFC5
Genomic Map