Assay Details
| Gene Symbol: MKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036674.2 | NP_001031751.1 | 3 - 4 | 382 | 74 | |
| NM_001125607.1 | NP_001119079.1 | 3 - 4 | 348 | 74 | ||
| NM_119127.4 | NP_194710.1 | 3 - 4 | 436 | 74 | ||
| GenBank mRNA | AB015313.1 | - | 3 - 4 | 300 | 74 | |
| AF385688.1 | - | 3 - 4 | 319 | 74 | ||
| AY078009.1 | - | 3 - 4 | 293 | 74 | ||
More Information
Set Membership: |
|

MKK2
Genomic Map