Assay Details
| Gene Symbol: MKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036674.2 | NP_001031751.1 | 4 - 5 | 604 | 102 | |
| NM_001125607.1 | NP_001119079.1 | 4 - 5 | 570 | 102 | ||
| NM_119127.4 | NP_194710.1 | 4 - 5 | 658 | 102 | ||
| GenBank mRNA | AB015313.1 | - | 4 - 5 | 522 | 102 | |
| AF385688.1 | - | 4 - 5 | 541 | 102 | ||
| AY078009.1 | - | 4 - 5 | 515 | 102 | ||
More Information
Set Membership: |
|

MKK2
Genomic Map