Assay Details
| Gene Symbol: MKK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036674.2 | NP_001031751.1 | 6 - 7 | 1020 | 96 | |
| NM_001125607.1 | NP_001119079.1 | 6 - 7 | 986 | 96 | ||
| NM_119127.4 | NP_194710.1 | 6 - 7 | 1074 | 96 | ||
| GenBank mRNA | AB015313.1 | - | 6 - 7 | 938 | 96 | |
| AF385688.1 | - | 6 - 7 | 957 | 96 | ||
| AY078009.1 | - | 6 - 7 | 931 | 96 | ||
More Information
Set Membership: |
|

MKK2
Genomic Map