Assay Details
| Gene Symbol: ACA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341980.1 | NP_001327988.1 | 18 - 19 | 2106 | 139 | |
| NM_119136.4 | NP_194719.1 | 18 - 19 | 1796 | 139 | ||
More Information
Set Membership: |
|

ACA10
Genomic Map