Assay Details
| Gene Symbol: ACA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341980.1 | NP_001327988.1 | 24 - 25 | 2747 | 95 | |
| NM_119136.4 | NP_194719.1 | 24 - 25 | 2437 | 95 | ||
More Information
Set Membership: |
|

ACA10
Genomic Map