Assay Details
| Gene Symbol: ACA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341980.1 | NP_001327988.1 | 26 - 27 | 3033 | 81 | |
| NM_119136.4 | NP_194719.1 | 26 - 27 | 2723 | 81 | ||
More Information
Set Membership: |
|

ACA10
Genomic Map