Assay Details
| Gene Symbol: ATR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342001.1 | NP_001328167.1 | 11 - 12 | 1549 | 95 | |
| NM_119167.4 | NP_194750.1 | 11 - 12 | 1519 | 95 | ||
| GenBank mRNA | AF325101.1 | - | 11 - 12 | 1344 | 95 | |
More Information
Set Membership: |
|

ATR2
Genomic Map