Assay Details
| Gene Symbol: ATR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342001.1 | NP_001328167.1 | 14 - 15 | 2022 | 75 | |
| NM_119167.4 | NP_194750.1 | 14 - 15 | 1992 | 75 | ||
| NM_179141.2 | NP_849472.2 | 14 - 15 | 1956 | 75 | ||
| GenBank mRNA | AF325101.1 | - | 14 - 15 | 1817 | 75 | |
More Information
Set Membership: |
|

ATR2
Genomic Map