Assay Details
| Gene Symbol: BRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119207.3 | NP_194790.1 | 7 - 8 | 1238 | 56 | |
| GenBank mRNA | BT002334.1 | - | 7 - 8 | 1148 | 56 | |
More Information
Set Membership: |
|

BRS1
Genomic Map