Assay Details
| Gene Symbol: AT4G30780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119224.5 | NP_194807.1 | 3 - 4 | 868 | 80 | |
| GenBank mRNA | BT004291.1 | - | 3 - 4 | 792 | 80 | |
| BT005518.1 | - | 3 - 4 | 631 | 80 | ||
More Information
Set Membership: |
|

AT4G30780
Genomic Map