Assay Details
| Gene Symbol: AT4G30990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342048.1 | NP_001320096.1 | 16 - 17 | 2574 | 74 | |
| NM_001342049.1 | NP_001328345.1 | 16 - 17 | 2577 | 74 | ||
| NM_119247.7 | NP_194828.7 | 16 - 17 | 2577 | 74 | ||
More Information
Set Membership: |
|

AT4G30990
Genomic Map