Assay Details
| Gene Symbol: SKB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119262.6 | NP_194841.2 | 20 - 21 | 1788 | 99 | |
| NM_202918.2 | NP_974647.1 | 20 - 21 | 1729 | 99 | ||
More Information
Set Membership: |
|

SKB1
Genomic Map