Assay Details
| Gene Symbol: SKB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119262.6 | NP_194841.2 | 2 - 3 | 377 | 86 | |
| NM_202918.2 | NP_974647.1 | 2 - 3 | 318 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SKB1| Gene Symbol: SKB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119262.6 | NP_194841.2 | 2 - 3 | 377 | 86 | |
| NM_202918.2 | NP_974647.1 | 2 - 3 | 318 | 86 | ||
Set Membership: |
|