Assay Details
| Gene Symbol: PBA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036682.1 | NP_001031759.1 | 2 - 3 | 345 | 105 | |
| NM_001203950.1 | NP_001190879.1 | 3 - 4 | 237 | 105 | ||
| NM_119279.5 | NP_194858.1 | 2 - 3 | 364 | 105 | ||
| GenBank mRNA | AF043529.1 | - | 2 - 3 | 333 | 105 | |
| AY052354.1 | - | 2 - 3 | 345 | 105 | ||
| AY058240.1 | - | 2 - 3 | 181 | 105 | ||
| Y13694.1 | - | 252 | 105 | |||
More Information
Set Membership: |
|

PBA1
Genomic Map