Assay Details
| Gene Symbol: PBA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036682.1 | NP_001031759.1 | 7 - 8 | 731 | 79 | |
| NM_001203950.1 | NP_001190879.1 | 8 - 9 | 620 | 79 | ||
| NM_119279.5 | NP_194858.1 | 7 - 8 | 747 | 79 | ||
| GenBank mRNA | AF043529.1 | - | 7 - 8 | 716 | 79 | |
| AY052354.1 | - | 7 - 8 | 728 | 79 | ||
| AY058240.1 | - | 7 - 8 | 564 | 79 | ||
| Y13694.1 | - | 635 | 79 | |||
More Information
Set Membership: |
|

PBA1
Genomic Map