Assay Details
| Gene Symbol: MRE11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345081.1 | NP_001330550.1 | 18 - 19 | 1818 | 61 | |
| NM_124806.2 | NP_200237.1 | 20 - 21 | 2004 | 61 | ||
| GenBank mRNA | AJ243822.1 | - | 20 - 21 | 1973 | 61 | |
More Information
Set Membership: |
|

MRE11
Genomic Map