Assay Details
| Gene Symbol: BBX29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124827.5 | NP_200258.2 | 1 - 2 | 721 | 107 | |
| GenBank mRNA | AK221519.1 | - | 1 - 2 | 654 | 107 | |
| BT012358.1 | - | 1 - 2 | 497 | 107 | ||
More Information
Set Membership: |
|

BBX29
Genomic Map