Assay Details
| Gene Symbol: BGLU41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345090.1 | NP_001330204.1 | 4 - 5 | 350 | 150 | |
| NM_124837.3 | NP_200268.3 | 4 - 5 | 350 | 150 | ||
More Information
Set Membership: |
|

BGLU41
Genomic Map