Assay Details
| Gene Symbol: GPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124861.5 | NP_568812.1 | 1 - 2 | 765 | 73 | |
| GenBank mRNA | AF233658.1 | - | 1 - 2 | 562 | 73 | |
| AK226479.1 | - | 1 - 2 | 621 | 73 | ||
| AY057679.1 | - | 1 - 2 | 556 | 73 | ||
| BT002212.1 | - | 1 - 2 | 491 | 73 | ||
More Information
Set Membership: |
|

GPT1
Genomic Map