Assay Details
| Gene Symbol: GPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124861.5 | NP_568812.1 | 2 - 3 | 997 | 74 | |
| GenBank mRNA | AF233658.1 | - | 2 - 3 | 794 | 74 | |
| AK226479.1 | - | 2 - 3 | 853 | 74 | ||
| AY057679.1 | - | 2 - 3 | 788 | 74 | ||
| BT002212.1 | - | 2 - 3 | 723 | 74 | ||
More Information
Set Membership: |
|

GPT1
Genomic Map