Assay Details
| Gene Symbol: AT5G54980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124880.3 | NP_200309.1 | 1 - 2 | 381 | 73 | |
| GenBank mRNA | BT002961.1 | - | 1 - 2 | 316 | 73 | |
| BT004438.1 | - | 1 - 2 | 216 | 73 | ||
More Information
Set Membership: |
|

AT5G54980
Genomic Map