Assay Details
| Gene Symbol: SUMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345119.1 | NP_001330302.1 | 2 - 3 | 359 | 107 | |
| NM_124899.4 | NP_200328.1 | 2 - 3 | 353 | 107 | ||
| GenBank mRNA | AF510521.1 | - | 2 - 3 | 219 | 107 | |
| BT024755.1 | - | 2 - 3 | 219 | 107 | ||
More Information
Set Membership: |
|

SUMO3
Genomic Map