Assay Details
| Gene Symbol: RAN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124901.5 | NP_200330.1 | 7 - 8 | 825 | 84 | |
| GenBank mRNA | AY042796.1 | - | 7 - 8 | 655 | 84 | |
| AY050317.1 | - | 7 - 8 | 688 | 84 | ||
| AY116939.1 | - | 6 - 7 | 603 | 84 | ||
| U73810.1 | - | 6 - 7 | 603 | 84 | ||
More Information
Set Membership: |
|

RAN3
Genomic Map