Assay Details
| Gene Symbol: AT5G55220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124904.5 | NP_200333.2 | 9 - 10 | 1454 | 99 | |
| GenBank mRNA | AY074845.1 | - | 9 - 10 | 1295 | 99 | |
| BT002296.1 | - | 9 - 10 | 1234 | 99 | ||
More Information
Set Membership: |
|

AT5G55220
Genomic Map