Assay Details
| Gene Symbol: FTSZ1-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124910.6 | NP_200339.1 | 4 - 5 | 913 | 77 | |
| GenBank mRNA | AY034992.1 | - | 4 - 5 | 836 | 77 | |
| AY113896.1 | - | 4 - 5 | 791 | 77 | ||
More Information
Set Membership: |
|

FTSZ1-1
Genomic Map