Assay Details
| Gene Symbol: TOP1ALPHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203614.1 | NP_001190543.1 | 12 - 13 | 2461 | 93 | |
| NM_001203615.1 | NP_001190544.1 | 6 - 7 | 1398 | 93 | ||
| NM_124912.5 | NP_200341.1 | 12 - 13 | 2473 | 93 | ||
| GenBank mRNA | X57544.1 | - | 2398 | 93 | ||
More Information
Set Membership: |
|

TOP1ALPHA
Genomic Map