Assay Details
| Gene Symbol: TOP1ALPHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203614.1 | NP_001190543.1 | 3 - 4 | 1559 | 101 | |
| NM_124912.5 | NP_200341.1 | 3 - 4 | 1571 | 101 | ||
| GenBank mRNA | X57544.1 | - | 1496 | 101 | ||
More Information
Set Membership: |
|

TOP1ALPHA
Genomic Map