Assay Details
| Gene Symbol: TOP1BETA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124913.4 | NP_200342.1 | 4 - 5 | 1644 | 92 | |
| GenBank mRNA | AK226832.1 | - | 4 - 5 | 1560 | 92 | |
More Information
Set Membership: |
|

TOP1BETA
Genomic Map