Assay Details
| Gene Symbol: BAM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345148.1 | NP_001318804.1 | 4 - 5 | 972 | 99 | |
| NM_124952.5 | NP_568829.2 | 4 - 5 | 955 | 99 | ||
More Information
Set Membership: |
|

BAM4
Genomic Map