Assay Details
| Gene Symbol: NMNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203622.1 | NP_001190551.1 | 1 - 2 | 163 | 73 | |
| NM_001345151.1 | NP_001329970.1 | 1 - 2 | 163 | 73 | ||
| NM_124963.5 | NP_200392.3 | 1 - 2 | 221 | 73 | ||
More Information
Set Membership: |
|

NMNAT
Genomic Map