Assay Details
| Gene Symbol: NMNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203622.1 | NP_001190551.1 | 6 - 7 | 642 | 70 | |
| NM_124963.5 | NP_200392.3 | 6 - 7 | 622 | 70 | ||
More Information
Set Membership: |
|

NMNAT
Genomic Map