Assay Details
| Gene Symbol: NMNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203622.1 | NP_001190551.1 | 7 - 8 | 744 | 137 | |
| NM_001345151.1 | NP_001329970.1 | 6 - 7 | 835 | 137 | ||
| NM_124963.5 | NP_200392.3 | 7 - 8 | 724 | 137 | ||
More Information
Set Membership: |
|

NMNAT
Genomic Map