Assay Details
| Gene Symbol: CBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345176.1 | NP_001332655.1 | 7 - 8 | 952 | 112 | |
| NM_124981.4 | NP_200410.1 | 7 - 8 | 952 | 112 | ||
| GenBank mRNA | AF076252.1 | - | 6 - 7 | 490 | 112 | |
| AY052304.1 | - | 6 - 7 | 535 | 112 | ||
| AY139808.1 | - | 5 - 6 | 426 | 112 | ||
More Information
Set Membership: |
|

CBL2
Genomic Map