Assay Details
| Gene Symbol: CRK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341575.1 | NP_001329890.1 | 2 - 3 | 1045 | 79 | |
| NM_179094.3 | NP_849425.1 | 2 - 3 | 1039 | 79 | ||
| GenBank mRNA | AY142496.1 | - | 2 - 3 | 959 | 79 | |
More Information
Set Membership: |
|

CRK5
Genomic Map