Assay Details
| Gene Symbol: NUDT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036636.2 | NP_001031713.1 | 2 - 3 | 404 | 76 | |
| NM_001341732.1 | NP_001329739.1 | 2 - 3 | 368 | 76 | ||
| NM_001341733.1 | NP_001329738.1 | 2 - 3 | 340 | 76 | ||
| NM_001341734.1 | NP_001329741.1 | 2 - 3 | 340 | 76 | ||
| NM_001341735.1 | NP_001329740.1 | 1 - 2 | 498 | 76 | ||
| NM_001341739.1 | NP_001329745.1 | 2 - 3 | 368 | 76 | ||
| NM_001341740.1 | NP_001329744.1 | 2 - 3 | 368 | 76 | ||
| NM_001341741.1 | NP_001329748.1 | 2 - 3 | 368 | 76 | ||
| NM_001341742.1 | NP_001329747.1 | 2 - 3 | 368 | 76 | ||
| NM_179112.3 | NP_849443.2 | 2 - 3 | 368 | 76 | ||
| GenBank mRNA | BT010765.1 | - | 2 - 3 | 332 | 76 | |
| BT010987.1 | - | 2 - 3 | 240 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001341735.1 transcripts.
Set Membership: |
|

NUDT10
Genomic Map